1-Fluor-4-ethinylbenzol
Lieferant: FLUOROCHEM
Synonyme:
(4-Fluorphenyl)acetylen, 1-Ethinyl-4-fluorbenzol, (4-Fluorphenyl)ethin
008808-1G
FLUO008808-1GEA
60.9
EUR
FLUO008808-1G
1-Fluor-4-ethinylbenzol
1-Fluor-4-ethinylbenzol
Formel:
C₈H₅F Molecular Weight: 120.13 g/mol Flammpunkt: <61 °C |
MDL:
MFCD00168823 CAS-Nummer: 766-98-3 UN: 1325 ADR: 4.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...