4-Brombenzylalkohol

Lieferant: FLUOROCHEM

Synonyme: (4-Bromophenyl)methanol

008822-10G
FLUO008822-10GEA 0 EUR
FLUO008822-10G
4-Brombenzylalkohol
4-Brombenzylalkohol
Formel: C₇H₇BrO
Molecular Weight: 187,04 g/mol
Siedepunkt: 268 °C (1013 hPa)
Schmelzpunkt: 77…78 °C
MDL: MFCD00004650
CAS-Nummer: 873-75-6
EINECS: 212-851-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR