4-Chlorbenzylalkohol

Lieferant: FLUOROCHEM

Synonyme: (4-Chlorophenyl)methanol

008823-25G
FLUO008823-25GEA 36.9 EUR
FLUO008823-25G
4-Chlorbenzylalkohol
4-Chlorbenzylalkohol
Formel: C₇H₇ClO
Molecular Weight: 142,58 g/mol
Siedepunkt: 234 °C (1013 hPa)
Schmelzpunkt: 67…70 °C
MDL: MFCD00004652
CAS-Nummer: 873-76-7
EINECS: 212-852-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR