3,4-Dichlorbenzylalkohol
Lieferant: FLUOROCHEM
Synonyme:
(3,4-Dichlorophenyl)methanol
008824-10G
FLUO008824-10GEA
36.2
EUR
FLUO008824-10G
3,4-Dichlorbenzylalkohol
3,4-Dichlorbenzylalkohol
Formel:
C₇H₆Cl₂O Molecular Weight: 177,03 g/mol Siedepunkt: 271 °C (1013 hPa) Schmelzpunkt: 32…34 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00004633 CAS-Nummer: 1805-32-9 EINECS: 217-299-0 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...