3,4,5-Trimethoxybenzylalkohol
Lieferant: FLUOROCHEM
Synonyme:
(3,4,5-Trimethoxyphenyl)methanol
008831-10G
FLUO008831-10GEA
36.9
EUR
FLUO008831-10G
3,4,5-Trimethoxybenzylalkohol
3,4,5-Trimethoxybenzylalkohol
Formel:
C₁₀H₁₄O₄ Molecular Weight: 198,22 g/mol Siedepunkt: 350 °C (1013 hPa) Schmelzpunkt: 36…38 °C Dichte: 1,1427 g/cm³ (20 °C) |
MDL:
MFCD00060356 CAS-Nummer: 3840-31-1 EINECS: 223-331-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...