(±)-1-Indanol

Lieferant: FLUOROCHEM

Synonyme: (±)-1-Hydroxyindan

008845-25G
FLUO008845-25GEA 101 EUR
FLUO008845-25G
(±)-1-Indanol
(±)-1-Indanol
Formel: C₉H₁₀O
Molecular Weight: 134,18 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00003797
CAS-Nummer: 6351-10-6
EINECS: 228-755-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR