9-Hydroxyfluoren
Lieferant: FLUOROCHEM
Synonyme:
9-Fluorenol
008846-5G
008846-25G
FLUO008846-5GEA
0
EUR
FLUO008846-5G
FLUO008846-25G
9-Hydroxyfluoren
9-Hydroxyfluoren
Formel:
C₁₃H₁₀O Molecular Weight: 182.22 g/mol Siedepunkt: 320 Schmelzpunkt: 152…157 °C Dichte: 1.151 g/cm³ (20 °C) |
MDL:
MFCD00001135 CAS-Nummer: 1689-64-1 EINECS: 216-879-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...