2-Brom-4,6-dichloranilin
Lieferant: FLUOROCHEM
|
Achtung
|
008852-10G
FLUO008852-10GEA
66.8
EUR
FLUO008852-10G
2-Brom-4,6-dichloranilin
2-Brom-4,6-dichloranilin
Formel:
C₆H₄BrCl₂N Molecular Weight: 240,91 g/mol Siedepunkt: 273 °C (1013 hPa) Schmelzpunkt: 77…78 °C |
MDL:
MFCD00040936 CAS-Nummer: 697-86-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...