2,5-Difluorphenylisothiocyanat
Lieferant: FLUOROCHEM
|
Gefahr
|
Synonyme:
1,4-Difluoro-2-isothiocyanatobenzene
008857-5G
008857-25G
FLUO008857-5GEA
219
EUR
FLUO008857-5G
FLUO008857-25G
2,5-Difluorphenylisothiocyanat
2,5-Difluorphenylisothiocyanat
Formel:
C₇H₃F₂NS Siedepunkt: 111…112 °C (35 mmHg) Dichte: 1,338 g/cm³ (20 °C) |
MDL:
MFCD00041046 CAS-Nummer: 206559-57-1 UN: 3334 ADR: 9, |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...