6-Amino-1,4-benzodioxan

Lieferant: FLUOROCHEM

Synonyme: 2,3-Dihydrobenzo[b][1,4]dioxin-6-amine, 1,4-Benzodioxan-6-amine

008859-25G
FLUO008859-25GEA 266 EUR
FLUO008859-25G
6-Amino-1,4-benzodioxan
6-Amino-1,4-benzodioxan
Formel: C₈H₉NO₂
Molecular Weight: 151,17 g/mol
MDL: MFCD00006824
CAS-Nummer: 22013-33-8
EINECS: 244-718-4
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR