4-Hydroxybenzylamin Hydrobromid

Lieferant: FLUOROCHEM

Synonyme: 4-(Aminomethyl)phenol hydrobromide

008872-1G 008872-5G
FLUO008872-1GEA 55.4 EUR
FLUO008872-1G FLUO008872-5G
4-Hydroxybenzylamin Hydrobromid
4-Hydroxybenzylamin Hydrobromid
Formel: C₇H₁₀BrNO
Molecular Weight: 204.0644 g/mol
MDL: MFCD02258769
CAS-Nummer: 90430-14-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR