3,4-Dimethoxybenzylamin
Lieferant: FLUOROCHEM
008881-5G
FLUO008881-5GEA
90.9
EUR
FLUO008881-5G
3,4-Dimethoxybenzylamin
3,4-Dimethoxybenzylamin
Formel:
C₉H₁₃NO₂ Siedepunkt: 154…158 °C (12 mmHg) Dichte: 1,109 g/cm³ (20 °C) |
MDL:
MFCD00008116 CAS-Nummer: 5763-61-1 EINECS: 227-287-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...