3,4,5-Trimethoxybenzylamin
Lieferant: FLUOROCHEM
008882-1G
FLUO008882-1GEA
34.1
EUR
FLUO008882-1G
3,4,5-Trimethoxybenzylamin
3,4,5-Trimethoxybenzylamin
Formel:
C₁₀H₁₅NO₃ Molecular Weight: 197,23 g/mol Siedepunkt: 106 °C (0,1 mmHg) Dichte: 1,155 g/cm³ (20 °C) |
MDL:
MFCD00017150 CAS-Nummer: 18638-99-8 EINECS: 242-468-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...