3,4-Dimethoxyphenethylamin

Lieferant: FLUOROCHEM

Synonyme: 2-(3,4-Dimethoxyphenyl)ethylamine

008886-25G
FLUO008886-25GEA 0 EUR
FLUO008886-25G
3,4-Dimethoxyphenethylamin
3,4-Dimethoxyphenethylamin
Formel: C₁₀H₁₅NO₂
Molecular Weight: 181,23 g/mol
MDL: MFCD00008188
CAS-Nummer: 120-20-7
EINECS: 204-376-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR