3-Phenylpropylamin
Lieferant: FLUOROCHEM
Synonyme:
1-Amino-3-phenylpropane, 1-Amino-3-phenylpropan
008890-10G
FLUO008890-10GEA
0
EUR
FLUO008890-10G
3-Phenylpropylamin
3-Phenylpropylamin
Formel:
C₉H₁₃N Molecular Weight: 135,21 g/mol Siedepunkt: 221 °C (1013 hPa) Dichte: 0,951 g/cm³ (15 °C) |
MDL:
MFCD00008224 CAS-Nummer: 2038-57-5 EINECS: 218-012-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...