Ethyl-2-isothiocyanatobenzoat
Lieferant: FLUOROCHEM
Synonyme:
o-Ethoxycarbonylphenylisothiocyanat
008927-5G
008927-25G
008927-50G
FLUO008927-5GEA
134
EUR
FLUO008927-5G
FLUO008927-25G
FLUO008927-50G
Ethyl-2-isothiocyanatobenzoat
Ethyl-2-isothiocyanatobenzoat
Formel:
C₁₀H₉NO₂S Siedepunkt: 150…151 °C (1 mmHg) Dichte: 1,201 g/cm³ (20 °C) |
MDL:
MFCD00041063 CAS-Nummer: 99960-09-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...