Methyl-2-isothiocyanatobenzoat

Lieferant: FLUOROCHEM

Synonyme: o-Methoxycarbonylphenylisothiocyanat

008999-5G 008999-25G
FLUO008999-5GEA 137 EUR
FLUO008999-5G FLUO008999-25G
Methyl-2-isothiocyanatobenzoat
Methyl-2-isothiocyanatobenzoat
Formel: C₉H₇NO₂S
Molecular Weight: 193.23 g/mol
Siedepunkt: 104
Dichte: 1.25 g/cm³ (20 °C)
MDL: MFCD00041062
CAS-Nummer: 16024-82-1
EINECS: 240-164-2
UN: 2923
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR