(Bis(trifluoracetoxy)iod)pentafluorbenzol
Lieferant: FLUOROCHEM
009039-1G
FLUO009039-1GEA
227
EUR
FLUO009039-1G
(Bis(trifluoracetoxy)iod)pentafluorbenzol
(Bis(trifluoracetoxy)iod)pentafluorbenzol
Formel:
C₁₀F₁₁IO₄ Molecular Weight: 519,99 g/mol |
MDL:
MFCD00191612 CAS-Nummer: 14353-88-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...