3,3'-Bis(trifluormethyl)benzophenon

Lieferant: FLUOROCHEM

Synonyme: Bis(3-(trifluoromethyl)phenyl)methanone

009045-25G
FLUO009045-25GEA 494 EUR
FLUO009045-25G
3,3'-Bis(trifluormethyl)benzophenon
3,3'-Bis(trifluormethyl)benzophenon
Formel: C₁₅H₈F₆O
Molecular Weight: 318,22 g/mol
MDL: MFCD00000390
CAS-Nummer: 1868-00-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR