1-(3,5-Bis(trifluormethyl)phenyl)thioharnstoff

Lieferant: FLUOROCHEM

Synonyme: N-[3,5-Bis(trifluoromethyl)phenyl]thiourea

009050-10G
FLUO009050-10GEA 0 EUR
FLUO009050-10G
1-(3,5-Bis(trifluormethyl)phenyl)thioharnstoff
1-(3,5-Bis(trifluormethyl)phenyl)thioharnstoff
Formel: C₉H₆F₆N₂S
Molecular Weight: 288,22 g/mol
MDL: MFCD00041172
CAS-Nummer: 175277-17-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR