1-tert-Butyl-4-ethinylbenzol

Lieferant: FLUOROCHEM

Synonyme: 1-Ethinyl-4-tert-butylbenzol, (4-tert-Butylphenyl)acetylen, 4-Ethinyl-1-tert-butylbenzol, (4-tert-Butylphenyl)ethin, 4-tert-Butyl-1-ethinylbenzol

CFF875. 009071-10G 009071-50G
CHMPCFF875.EA 0 EUR
CHMPCFF875. FLUO009071-10G FLUO009071-50G
1-tert-Butyl-4-ethinylbenzol
1-tert-Butyl-4-ethinylbenzol
Formel: C₁₂H₁₄
MDL: MFCD00190197
CAS-Nummer: 772-38-3
UN: 3082
ADR: 9,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR