3-Butin-2-on
Lieferant: FLUOROCHEM
009078-5G
009078-25G
FLUO009078-5GEA
148
EUR
FLUO009078-5G
FLUO009078-25G
3-Butin-2-on
3-Butin-2-on
Formel:
C₄H₄O Siedepunkt: 85 °C (1013 hPa) Dichte: 0,886 g/cm³ (15 °C) |
MDL:
MFCD00008775 CAS-Nummer: 1423-60-5 EINECS: 215-834-2 UN: 1992 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...