2-Chlor-3-ethylbenzoxazoliumtetrafluoroborat

Lieferant: FLUOROCHEM

009081-5G
FLUO009081-5GEA 0 EUR
FLUO009081-5G
2-Chlor-3-ethylbenzoxazoliumtetrafluoroborat
2-Chloro-3-ethylbenzoxazolium tetrafluoroborate
Formel: C₉H₉BClF₄NO
MDL: MFCD00011943
CAS-Nummer: 63212-53-3
EINECS: 263-998-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR