3-Chlor-4-fluortoluol
Lieferant: FLUOROCHEM
|
Gefahr
|
Synonyme:
1-Chlor-2-fluor-5-methylbenzol
009090-10G
009090-25G
009090-5G
FLUO009090-10GEA
67.1
EUR
FLUO009090-10G
FLUO009090-25G
FLUO009090-5G
3-Chlor-4-fluortoluol
3-Chlor-4-fluortoluol
Formel:
C₇H₆ClF Molecular Weight: 144.58 g/mol |
MDL:
MFCD00079730 CAS-Nummer: 1513-25-3 UN: 1993 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...