5-Fluor-2'-deoxyuridin (Floxuridin)
Lieferant: FLUOROCHEM
Synonyme:
5-Fluoro-1-((2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidine-2,4(1H,3H)-dione, 5-Fluoro-2'-deoxyuridine, Floxuridine
009121-500MG
FLUO009121-500MGEA
0
EUR
FLUO009121-500MG
5-Fluor-2'-deoxyuridin (Floxuridin)
5-Fluor-2'-deoxyuridin (Floxuridin)
Formel:
C₉H₁₁FN₂O₅ Molecular Weight: 246,19 g/mol |
MDL:
MFCD00006530 CAS-Nummer: 50-91-9 EINECS: 200-072-5 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...