2,5-Dibrom-α,α,α-trifluortoluol

Lieferant: FLUOROCHEM

Synonyme: 1,4-Dibromo-2-(trifluoromethyl)benzene, 2,5-Dibromobenzotrifluoride

009124-5G 009124-25G
FLUO009124-5GEA 48.8 EUR
FLUO009124-5G FLUO009124-25G
2,5-Dibrom-α,α,α-trifluortoluol
2,5-Dibrom-α,α,α-trifluortoluol
Formel: C₇H₃Br₂F₃
Molecular Weight: 303.9 g/mol
MDL: MFCD00013554
CAS-Nummer: 7657-09-2
EINECS: 231-620-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR