2,5-Dibrom-α,α,α-trifluortoluol
Lieferant: FLUOROCHEM
Synonyme:
1,4-Dibromo-2-(trifluoromethyl)benzene, 2,5-Dibromobenzotrifluoride
009124-5G
009124-25G
FLUO009124-5GEA
48.8
EUR
FLUO009124-5G
FLUO009124-25G
2,5-Dibrom-α,α,α-trifluortoluol
2,5-Dibrom-α,α,α-trifluortoluol
Formel:
C₇H₃Br₂F₃ Molecular Weight: 303.9 g/mol |
MDL:
MFCD00013554 CAS-Nummer: 7657-09-2 EINECS: 231-620-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...