2,6-Di-tert-butyl-4-methylpyryliumtrifluormethansulfonat
Lieferant: FLUOROCHEM
009126-5G
FLUO009126-5GEA
272
EUR
FLUO009126-5G
2,6-Di-tert-butyl-4-methylpyryliumtrifluormethansulfonat
2,6-Di-tert-butyl-4-methylpyryliumtrifluormethansulfonat
Formel:
C₁₅H₂₃F₃O₄S |
MDL:
MFCD00229854 CAS-Nummer: 59643-43-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...