1-Ethyl-3-methylimidazoliumhexafluorophosphat
Lieferant: FLUOROCHEM
Synonyme:
1-Ethyl-3-methyl-1H-imidazol-3-ium hexafluorophosphate(V)
009179-5G
FLUO009179-5GEA
0
EUR
FLUO009179-5G
1-Ethyl-3-methylimidazoliumhexafluorophosphat
1-Ethyl-3-methylimidazoliumhexafluorophosphate
Formel:
C₆H₁₁F₆N₂P Molecular Weight: 256,13 g/mol |
MDL:
MFCD00216703 CAS-Nummer: 155371-19-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...