1-Ethyl-3-methylimidazoliumtrifluormethansulfonat

Lieferant: FLUOROCHEM

Synonyme: 1-Ethyl-3-methylimidazolium trifluoromethanesulphonate

009181-5G
FLUO009181-5GEA 228 EUR
FLUO009181-5G
1-Ethyl-3-methylimidazoliumtrifluormethansulfonat
1-Ethyl-3-methylimidazoliumtrifluoromethanesulphonate
Formel: C₇H₁₁F₃N₂O₃S
MDL: MFCD00210008
CAS-Nummer: 145022-44-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR