2-Fluorbenzalchlorid
Lieferant: FLUOROCHEM
Synonyme:
α,α-Dichloro-2-fluortoluene
009190-10G
FLUO009190-10GEA
0
EUR
FLUO009190-10G
2-Fluorbenzalchlorid
2-Fluorbenzalchlorid
Formel:
C₇H₅Cl₂F Molecular Weight: 179,02 g/mol Siedepunkt: 71,6 °C (13 mmHg) Dichte: 1,3094 g/cm³ (18.7 °C) |
MDL:
MFCD00043797 CAS-Nummer: 320-65-0 EINECS: 206-279-7 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...