3-Fluorbenzalchlorid
Lieferant: FLUOROCHEM
|
Gefahr
|
Synonyme:
α,α-Dichlor-3-fluortoluol
009191-10G
FLUO009191-10GEA
0
EUR
FLUO009191-10G
3-Fluorbenzalchlorid
3-Fluorbenzalchlorid
Formel:
C₇H₅Cl₂F Molecular Weight: 179,02 g/mol Siedepunkt: 202…204 °C (1013 hPa) Dichte: 1,36 g/cm³ (16 °C) |
MDL:
MFCD00037112 CAS-Nummer: 402-64-2 EINECS: 206-952-5 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...