4-Fluor-3-iodtoluol

Lieferant: FLUOROCHEM

009207-10G 009207-25G 009207-5G
FLUO009207-10GEA 55.4 EUR
FLUO009207-10G FLUO009207-25G FLUO009207-5G
4-Fluor-3-iodtoluol
4-Fluor-3-iodtoluol
Formel: C₇H₆FI
Molecular Weight: 236.03 g/mol
MDL: MFCD00079759
CAS-Nummer: 452-82-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR