2-Fluor-5-nitrobenzoesäure

Lieferant: CHEMPUR
Achtung

CFF1134. FL9222.10 009222-25G 009222-5G 009222-100G
CHMPCFF1134.EA 0 EUR
CHMPCFF1134. CHMPFL9222.10 FLUO009222-25G FLUO009222-5G FLUO009222-100G
2-Fluor-5-nitrobenzoesäure
2-Fluor-5-nitrobenzoesäure
Formel: C₇H₄FNO₄
MDL: MFCD00134238
CAS-Nummer: 7304-32-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR