2-Hydroxynicotinsäure

Lieferant: FLUOROCHEM

Synonyme: 2-Hydroxypyridin-3-carbonsäure, 1,2-Dihydro-2-oxonicotinsäure, 2-Hydroxy-3-pyridincarbonsäure

009264-100G 009264-500G
FLUO009264-100GEA 119 EUR
FLUO009264-100G FLUO009264-500G
2-Hydroxynicotinsäure
2-Hydroxynicotinsäure
Formel: C₆H₅NO₃
MDL: MFCD00010100
CAS-Nummer: 609-71-2
EINECS: 210-198-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR