(S)-(-)-α-Methoxy-α-(trifluormethyl)phenylessigsäure

Lieferant: FLUOROCHEM

Synonyme: (S)-(-)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetic acid, (2S)-(-)-2-Methoxy-2-phenyl-3,3,3-trifluoropropanoic acid

009274-1G 009274-5G
FLUO009274-1GEA 170 EUR
FLUO009274-1G FLUO009274-5G
(S)-(-)-α-Methoxy-α-(trifluormethyl)phenylessigsäure
(S)-(-)-α-Methoxy-α-(trifluormethyl)phenylessigsäure
Formel: C₁₀H₉F₃O₃
MDL: MFCD00064200
CAS-Nummer: 17257-71-5
EINECS: 241-292-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR