3-Methylpicolinsäure

Lieferant: FLUOROCHEM

Synonyme: 3-Methylpyridine-2-carboxylic acid

009287-1G
FLUO009287-1GEA 31.1 EUR
FLUO009287-1G
3-Methylpicolinsäure
3-Methylpicolinsäure
Formel: C₇H₇NO₂
Molecular Weight: 137.14 g/mol
MDL: MFCD00191553
CAS-Nummer: 4021-07-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR