N-Methyl-2,2,2-trifluoracetamid

Lieferant: FLUOROCHEM

Synonyme: N-Methyltrifluoroacetamide, 2,2,2-Trifluoro-N-methylacetamide

009293-25G
FLUO009293-25GEA 42.9 EUR
FLUO009293-25G
N-Methyl-2,2,2-trifluoracetamid
N-Methyl-2,2,2-trifluoracetamid
Formel: C₃H₄F₃NO
Molecular Weight: 127,07 g/mol
MDL: MFCD00009670
CAS-Nummer: 815-06-5
EINECS: 212-417-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR