4-Nitro-α,α,α-trifluortoluol
Lieferant: FLUOROCHEM
Synonyme:
1-Nitro-4-(trifluormethyl)benzol, 4-Nitrobenzotrifluorid
009303-25G
FLUO009303-25GEA
0
EUR
FLUO009303-25G
4-Nitro-α,α,α-trifluortoluol
4-Nitro-α,α,α-trifluortoluol
Formel:
C₇H₄F₃NO₂ Molecular Weight: 191,11 g/mol Siedepunkt: 204 °C (1013 hPa) Schmelzpunkt: 40…41 °C |
MDL:
MFCD00007358 CAS-Nummer: 402-54-0 EINECS: 206-948-3 UN: 3431 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...