1H,1H-Hentriacontafluor-1-hexadecanol
Lieferant: FLUOROCHEM
009325-10G
FLUO009325-10GEA
0
EUR
FLUO009325-10G
1H,1H-Hentriacontafluor-1-hexadecanol
1H,1H-Hentriacontafluor-1-hexadecanol
Formel:
C₁₆H₃F₃₁O |
MDL:
MFCD00236626 CAS-Nummer: 216144-94-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...