Scandium(III)trifluormethansulfonat
Lieferant: FLUOROCHEM
Synonyme:
Scandium(III) trifluoromethanesulphonate, Scandium triflate
009343-1G
009343-5G
FLUO009343-1GEA
34.1
EUR
FLUO009343-1G
FLUO009343-5G
Scandium(III)trifluormethansulfonat
Scandium(III)trifluormethansulfonat
Formel:
C₃F₉O₉S₃Sc |
MDL:
MFCD00192433 CAS-Nummer: 144026-79-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...