Tetraethylphosphoniumhexafluorophosphat

Lieferant: FLUOROCHEM

009345-5G
FLUO009345-5GEA 354 EUR
FLUO009345-5G
Tetraethylphosphoniumhexafluorophosphat
Tetraethylphosphonium hexafluorophosphate
Formel: C₈H₂₀F₆P₂
MDL: MFCD01631313
CAS-Nummer: 111928-07-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR