Tetraphenylphosphoniumbromid

Lieferant: FLUOROCHEM

009348-10G 009348-50G 009348-250G
FLUO009348-10GEA 0 EUR
FLUO009348-10G FLUO009348-50G FLUO009348-250G
Tetraphenylphosphoniumbromid
Tetraphenylphosphonium bromide
Formel: C₂₄H₂₀BrP
Schmelzpunkt: 299…300 °C
MDL: MFCD00011915
CAS-Nummer: 2751-90-8
EINECS: 220-393-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR