3-(Trifluormethoxy)zimtsäure

Lieferant: FLUOROCHEM

009359-25G 009359-5G
FLUO009359-25GEA 134 EUR
FLUO009359-25G FLUO009359-5G
3-(Trifluormethoxy)zimtsäure
3-(Trifluormethoxy)zimtsäure
Formel: C₁₀H₇F₃O₃
MDL: MFCD00066337
CAS-Nummer: 168833-80-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR