Ytterbium(III)trifluormethansulfonat

Lieferant: FLUOROCHEM

Synonyme: Ytterbium (III) triflate

009409-5G
FLUO009409-5GEA 121 EUR
FLUO009409-5G
Ytterbium(III)trifluormethansulfonat
Ytterbium(III)trifluormethansulfonat
Formel: C₃F₉O₉S₃Yb
MDL: MFCD06200261
CAS-Nummer: 54761-04-5
UN: 3261
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR