Tetrafluorterephthalonitril

Lieferant: FLUOROCHEM

Synonyme: Tetrafluor-1,4-benzoldicarbonitril, Tetrafluorbenzol-1,4-dicarbonitril, Tetrafluor-1,4-dicyanobenzol, Tetrafluorterephthalsäuredinitril, Tetrafluor-1,4-dicyanbenzol

009426-25G
FLUO009426-25GEA 116 EUR
FLUO009426-25G
Tetrafluorterephthalonitril
Tetrafluorterephthalonitril
Formel: C₈F₄N₂
Molecular Weight: 200,1 g/mol
MDL: MFCD00001776
CAS-Nummer: 1835-49-0
EINECS: 217-397-3
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR