2-Desoxy-2-fluor-D-glucose

Lieferant: FLUOROCHEM

Synonyme: 2-Fluoro-2-deoxy-D-glucose, 2-Deoxy-2-fluoro-D-glucopyranose, FDG

009448-25MG
FLUO009448-25MGEA 0 EUR
FLUO009448-25MG
2-Desoxy-2-fluor-D-glucose
2-Deoxy-2-fluoro-D-glucose
Formel: C₆H₁₁FO₅
Molecular Weight: 182,15 g/mol
MDL: MFCD00077527
CAS-Nummer: 29702-43-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR