5-Fluor-DL-tryptophan

Lieferant: FLUOROCHEM

Synonyme: 2-Amino-3-(5-fluoro-1H-indol-3-yl)propanoic acid, H-DL-Trp(5-F)-OH

009449-1G
FLUO009449-1GEA 350 EUR
FLUO009449-1G
5-Fluor-DL-tryptophan
5-Fluor-DL-tryptophan
Formel: C₁₁H₁₁FN₂O₂
Molecular Weight: 222,22 g/mol
Schmelzpunkt: 263…264 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00005649
CAS-Nummer: 154-08-5
EINECS: 205-822-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR