2-Brom-6-(trifluormethyl)pyridin

Lieferant: FLUOROCHEM

009467-1G 009467-5G
FLUO009467-1GEA 31.1 EUR
FLUO009467-1G FLUO009467-5G
2-Brom-6-(trifluormethyl)pyridin
2-Bromo-6-(trifluoromethyl)pyridine
Formel: C₆H₃BrF₃N
MDL: MFCD00153087
CAS-Nummer: 189278-27-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR