2-Chlorisonicotinonitril

Lieferant: FLUOROCHEM

CFF132.1 CFF132.2.5 CFF132.10 009501-10G 009501-50G
CHMPCFF132.1EA 242 EUR
CHMPCFF132.1 CHMPCFF132.2.5 CHMPCFF132.10 FLUO009501-10G FLUO009501-50G
2-Chlorisonicotinonitril
2-Chlorisonicotinonitril
Formel: C₆H₃ClN₂
MDL: MFCD00174318
CAS-Nummer: 33252-30-1
UN: 3439
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR